Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSSK6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567708
Description
Predicted to react with Mouse and Rat samples.
The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. TSSK 6 (testis-specific serine kinase 6), also known as SSTK, TSSK4, FKSG82 or CT72, is a 273 amino acid protein that contains one protein kinase domain and belongs to the Ser/Thr protein kinase family. Highly expressed in testis with lower expression in ovary, colon, thymus, small intestine and spleen, TSSK 6 catalyzes the ATP-dependent phosphorylation of proteins involved in sperm production and chromatin remodeling. TSSK 6 uses magnesium as a cofactor and is thought to be required for proper sperm development and function, as well as DNA condensation events. Defects in the gene encoding TSSK 6 are associated with male infertility characterized by low sperm count and decreased sperm motility.
Specifications
TSSK6 | |
Polyclonal | |
Unconjugated | |
TSSK6 | |
Cancer/testis antigen 72; CT72; FKSG82; RGD1559764; serine/threonine protein kinase SSTK; serine/threonine-protein kinase SSTK; Small serine/threonine kinase; SSTK; testis secretory sperm-binding protein Li 205a; testis specific serine kinase 6; testis-specific kinase 6; testis-specific serine kinase 6; testis-specific serine/threonine-protein kinase 6; TSK-6; TSSK4; TSSK6; TSSK-6 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
290670, 83983, 83984 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q925K9, Q9BXA6 | |
TSSK6 | |
Synthesized peptide derived from human TSSK6. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction