Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSSK6 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP286880

Catalog No. NB034502


Add to cart

Description

Description

TSSK6 Polyclonal specifically detects TSSK6 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

TSSK6
Unconjugated
PBS, 2% Sucrose
Cancer/testis antigen 72, CT72TSSK-6, EC 2.7.11.1, FLJ24002, serine/threonine protein kinase SSTK, Serine/threonine-protein kinase SSTK, Small serine/threonine kinase, SSTKTSK-6, Testis-specific kinase 6, testis-specific serine kinase 6, testis-specific serine/threonine-protein kinase 6, TSSK4
The immunogen is a synthetic peptide directed towards the middle region of mouse TSSK6
100 μg
Protein Kinase
Primary
83983
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Immunogen affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit