Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TST Recombinant Protein, Abnova
$420.00 - $645.00
Specifications
Cross Reactivity | Human |
---|---|
Format | Solution |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
For Use With (Application) | Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) |
Molecular Weight | 58.41 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-010-978
|
Abnova Corporation
H00007263P0110 |
10 ug |
Each for $420.00
|
|
89-010-979
|
Abnova Corporation
H00007263P0125 |
25 ug |
Each for $645.00
|
|
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Human | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
58.41 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Yes | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Solution | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
TST (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title