Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSV Value Lab Refrigerator/Freezer

Catalog No. p-9249007
Click to view available options
Capacity (English) Refrigerator:
5.1 cu. ft.
7.1 cu. ft.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Chill, freeze and retrieve samples safely, simply and economically with the Thermo Scientific™ TSV Value™ Refrigerator/Freezer.

  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clearn, durable finish
TRUSTED_SUSTAINABILITY

Specifications

Type General Purpose Combo
Capacity (English) Refrigerator 5.1 cu. ft.
Capacity (Metric) Refrigerator 144.4 L
Temperature Range (Freezer) -12°C to -20°C
Temperature Range (Refrigerator) 1°C to 12°C
Temperature (Setpoint) Adjustable
Door Style Hinged, Open Left to Right
No. of Doors 1
Inner Doors 1
Shelves 2 Adjustable
Door Shelves 2
Compressor Hermetic
Defrost Automatic (Refrigerator), Manual (Freezer)
Access Security None
Refrigerant R600a hydrocarbon refrigerant
Exterior Finish Enameled Steel
Insulation Cyclopentane
Interior ABS Plastic
Certifications/Compliance cULus
Casters 2 adjustable feet in front
Color White
Control Type Mechanical Dial
For Use With (Application) Indoor General Storage of Non-Critical Samples and Reagents
Min. Clearance 4 in. on all sides
Model TSV05CPSA
Typical Lifetime 10 Years
Warranty 12 Months
Dimensions (DxWxH) Exterior 23.5 x 23.63 x 33.5 in. (56.69 x 60.02 x 85.1cm)
Dimensions (D x W x H) Interior Refrigerator: 17 x 20.5 x 19.5 in.(43.2 x 52.1 x 49.5cm)Freezer: 11.5 x 19 x 5 in. (29.2 x 48.3 x 12.7cm)
Energy Usage .35 kWh/Day
Amperage 1.2A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115V 60Hz
Frequency 60 Hz
Shipping Weight (Metric) 46 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.