Learn More
Thermo Scientific™ TSX Series Ultra-Low Freezers - PROMO
Thermo Scientific™ TSX Series ultra-low freezers are designed to meet the highest sustainability, temperature management and reliability standards for -80°C and -70°C storage.
Specifications
Product Line TSX Series
Smart View Compatibility Smart-Vue Compatible
Warranty Warranty varies by country, please contact us for details
Defrost Manual
Depth (English) Interior 28.3 in.
View More Specs
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Products 8
Catalog Number Mfr. No. Holds Dimensions (DxWxH) Exterior Electrical Requirements Price Quantity & Availability  
Catalog Number Mfr. No. Holds Dimensions (DxWxH) Exterior Electrical Requirements Price Quantity & Availability  
TSX40086AQ2
View Documents
Thermo Scientific™
TSX40086AQ2
2 in. Boxes (400) 37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm) 115 V 60 Hz This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
TSX40086DQ2
View Documents
Thermo Scientific™
TSX40086DQ2
2 in. Boxes (400) 37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm) 208 to 230 V 60 Hz This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
TSX50086AQ2
View Documents
Thermo Scientific™
TSX50086AQ2
2 in. Boxes (500) 37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm) 115 V 60 Hz This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
TSX50086DQ2
View Documents
Thermo Scientific™
TSX50086DQ2
2 in. Boxes (500) 37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm) 208 to 230 V 60 Hz This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
TSX60086AQ2
View Documents
Thermo Scientific™
TSX60086AQ2
2 in. Boxes (600) 37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1cm) 115 V 60 Hz This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
TSX60086DQ2
View Documents
Thermo Scientific™
TSX60086DQ2
2 in. Boxes (600) 37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm) 208 to 230 V 60 Hz This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
TSX70086AQ2
View Documents
Thermo Scientific™
TSX70086AQ2
2 in. Boxes (700) 37.6 x 45.2 x 78 in. (95.5 x 114.8 x 198.1 cm) 115 V 60 Hz This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
TSX70086DQ2
View Documents
Thermo Scientific™
TSX70086DQ2
2 in. Boxes (700) 37.6 x 45.2 x 78 in. (95.5 x 114.8 x 198.1 cm) 208 to 230 V 60 Hz This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Performance
  • No need to worry about where your samples are stored within the freezer.  With peak variation less than 5°C, your samples are protected and safely stored throughout the entire cabinet space. *
  • Long warm-up time from -50°C to -80°C: 5.05 hours*
  • Quiet enough to reside directly in the laboratory, rather than in the hall. At just 45.5 dBA, the TSX Series’ sound level is comparable to a home refrigerator.*
  • V-drive’s adaptive control is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
Wireless Monitoring Solution
Thermo Scientific™ DeviceLink™ comes embedded in your ULT freezer  providing real-time monitoring of system status and sample temperature.
  • View critical lab equipment data and equipment notifications on your mobile and web applications.
    • Display temperature
    • Ambient temperature
    • Alarm and alerts notification
  • Access data anytime, anywhere—data and alarms from monitored lab equipment are securely stored on the Cloud and sent to the user via email and mobile-phone notifications
  • InstrumentConnect™ streamlined dashboard-style reporting reduces time spent interpreting data
  • Unlike traditional connectivity methods, DeviceLink™ operates on a client-cloud architecture, requiring minimal IT support
  • No need to install sensors—the only plug-in is the equipment data port to read the parameters directly from the factory-installed sensors
  • Applicable Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP
User Friendly
  • Newly re-designed touch-screen user interface with easy-to-use: set-up wizard, event log, temperature mapping and data downloads via a built-in USB port
  • Eliminate the need for a separate chart recorder with the touch-screen’s datalogging and temperature mapping features, all exportable via a built-in USB port
  • Ergonomic design including comfortable door handle, eye-level touch-screen and shelf-level placement that don’t require floor-level loading of racks
  • Four capacities (400, 500, 600 and 700 2-inch box) to accommodate a variety of storage needs and lab footprint specifications
  • Compatible with most racking systems including a full range of Thermo Scientific™ racks for boxes, microplates, Thermo Scientific™ Matrix™ and Nunc CryoBank™ tubes
  • Security features including alarm settings, padlock-compatible doors and optional access card entry
Sustainability
  • Natural refrigerants and water-blown foam insulation, making the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), the European Union’s F-Gas and other sustainability standards
  • Manufactured in an award-winning, zero-waste to landfill facility
  • Lower HVAC costs as a result of less heat emission into the environment
Footnotes:
* Typical freezer data based on internal testing with freezer setpoint at -80°C and ambient temperature at 20°C. Performance data varies across freezer size and electrical specifications.
Specifications
Specifications
TSX Series
Warranty varies by country, please contact us for details
28.3 in.
Touch-Screen LCD
51.2 in.
Smart-Vue Compatible
Manual
71.9 cm
78 in.
198.1 cm