Learn More
TSX ultra-low freezer package with racks, boxes and CO2 back-up
TSX ultra-low freezer package with racks, boxes and factory-installed CO2 back-up
Manufacturer:  Thermo Scientific TSX40086ARAKCO
 View more versions of this product
Catalog No. 09-313-876
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Not available outside North America
Description
Description
Thermo Scientific™ TSX Ultra-Low temperature freezer package with racks, boxes, and factory-installed CO2 back-up system.
Package includes:
· One TSX 500 box capacity ultra-low freezer, 208-230V, 60Hz
· Ten sliding drawer racks with 200 2-inch cryoboxes
· Factory-installed LN2 back-up
Warranty and Services
Warranty varies by country, please contact us for details
Specifications
Specifications
−50° to −86°C
Ultra Low Freezer, Upright
115V, 60Hz
400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers, and factory-installed CO2 back-up system