Learn More
TTC11 Mouse anti-Human, DyLight 650, Clone: 9810, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243628C
Catalog No. NBP243628C
Add to cart
Description
Description
TTC11 Monoclonal antibody specifically detects TTC11 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
TTC11
9810
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
FIS1
Full length human FIS1 Recombinant protein.
Protein G purified
Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors
Primary
51024
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human