Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC11 Mouse anti-Human, FITC, Clone: 4211, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243771F

Catalog No. NBP243771F


Add to cart

Description

Description

TTC11 Monoclonal antibody specifically detects TTC11 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

TTC11
4211
Immunocytochemistry/Immunofluorescence
Purified
FIS1
Full length human FIS1 Recombinant protein.
Protein A purified
Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors
Primary
51024
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit