Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC11 Rabbit anti-Human, Mouse, Rat, Clone: 6S1G4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | TTC11 |
---|---|
Clone | 6S1G4 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown |
Classification | Monoclonal |
Description
TTC11 Monoclonal antibody specifically detects TTC11 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDownSpecifications
TTC11 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-122 of human TTC11 (NP_057152.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
6S1G4 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
51024 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title