Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC15 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548569

Catalog No. PA548569

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

May be involved in endoplasmic reticulum to Golgi apparatus trafficking at a very early stage.
Specifications

Specifications

TTC15
Polyclonal
PBS with 0.09% sodium azide
Q8WVT3
Trafficking protein particle complex subunit 12, Tetratricopeptide repeat protein 15, TPR repeat protein 15, TTC-15, TRAPPC12, TTC15
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51112
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TRAPPC12
Liquid
TRAPPC12
KLH conjugated synthetic peptide between 579-608 amino acids from the C-terminal region of human TTC15.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit