Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC19 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19155220UL

 View more versions of this product

Catalog No. NBP19155220


Add to cart

Description

Description

TTC19 Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

TTC19
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
TTC19
Synthetic peptide directed towards the C terminal of human TTC19. Peptide sequence LAAILIHRERYTQAKEIYQEALKRAELKRDEVSVQHIREELAELSRKSRR.
Protein A purified
RUO
Primary
54902
Western Blot
Western Blot 1:1000
NP_082636
MGC138312, MGC19520, tetratricopeptide repeat domain 19,2010204O13Rik, tetratricopeptide repeat protein 19, mitochondrial, TPR repeat protein 19
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit