Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC23L Mouse anti-Human, Clone: 2A4, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $450.00
Specifications
Antigen | TTC23L |
---|---|
Clone | 2A4 |
Concentration | 1.0 mg/mL |
Applications | Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE187025
|
MilliporeSigma
MABE187025UG |
25 μg |
Each for $177.00
|
|
|||||
MABE1870100
|
MilliporeSigma
MABE1870100UG |
100 μg |
Each for $450.00
|
|
|||||
Description
TTC23L Monoclonal antibody specifically detects TTC23L in Human samples. It is validated for Immunofluorescence, Western Blotting.Specifications
TTC23L | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG1 κ | |
Protein G purified |
2A4 | |
Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Linear Peptide corresponding to 16 amino acids from the C-terminal region of human Tetratricopeptide repeat protein 23-like (TTC23L), isoform 1. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title