Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC27 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179659
Description
TTC27 Polyclonal specifically detects TTC27 in Human samples. It is validated for Western Blot.Specifications
TTC27 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ20272, tetratricopeptide repeat domain 27, tetratricopeptide repeat protein 27, TPR repeat protein 27 | |
Rabbit | |
97 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: yeast 85%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_060205 | |
TTC27 | |
Synthetic peptide directed towards the N terminal of human TTC27The immunogen for this antibody is TTC27. Peptide sequence KDQLDIAKDISQLQIDLTGALGKRTRFQENYVAQLILDVRREGDVLSNCE. | |
Affinity purified | |
RUO | |
55622 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction