Learn More
TTC31 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 216931AP
Catalog No. 50-173-3549
Add to cart
Specifications
Specifications
TTC31
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q49AM3
TPR repeat protein 31, TTC31
Rabbit
IgG
150 μL
-20°C
Primary
64427
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TTC31
Liquid
TTC31
TTC31 Fusion Protein Ag16281
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human