Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC32 Mouse anti-Human, Clone: OTI5B4, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA501339
Description
TTC32 is a protein coding gene.Specifications
TTC32 | |
Monoclonal | |
0.56 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q5I0X7 | |
tetratricopeptide repeat domain 32 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
130502 |
Flow Cytometry, Western Blot | |
OTI5B4 | |
Unconjugated | |
TTC32 | |
Liquid | |
TTC32 | |
Full length human recombit protein of human TTC32 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title