Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC32 Monoclonal Antibody (OTI5C12), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA525157
Description
TTC32 Monoclonal antibody specifically detects TTC32 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
TTC32 | |
Monoclonal | |
0.55 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5I0X7 | |
Ttc32 | |
Full length human recombinant protein of TTC32 produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI5C12 | |
Unconjugated | |
Ttc32 | |
1700013G20Rik; tetratricopeptide repeat domain 32; tetratricopeptide repeat protein 32; TPR repeat protein 32; TTC32 | |
Mouse | |
Affinity Chromatography | |
RUO | |
130502 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction