Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC33 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155426
Description
TTC33 Polyclonal specifically detects TTC33 in Human samples. It is validated for Western Blot.Specifications
TTC33 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
osmosis responsive factor, Osmosis-responsive factor, OSRF, tetratricopeptide repeat domain 33, tetratricopeptide repeat protein 33, TPR repeat protein 33 | |
Rabbit | |
29 kDa | |
100 μL | |
Signal Transduction | |
23548 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q6PID6 | |
TTC33 | |
Synthetic peptides corresponding to TTC33(tetratricopeptide repeat domain 33) The peptide sequence was selected from the N terminal of TTC33. Peptide sequence DEGNWLHAIKRRKEILLEGCAEKSKQLKDEGASLAENKRYREAIQKWDEA. | |
Affinity purified | |
RUO | |
Primary | |
Porcine: 86%; Guinea pig: 86%; Yeast: 82%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction