Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC33 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | TTC33 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP155426
|
Novus Biologicals
NBP155426 |
100 μL |
Each of 1 for $436.00
|
|
Description
TTC33 Polyclonal specifically detects TTC33 in Human samples. It is validated for Western Blot.Specifications
TTC33 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
osmosis responsive factor, Osmosis-responsive factor, OSRF, tetratricopeptide repeat domain 33, tetratricopeptide repeat protein 33, TPR repeat protein 33 | |
TTC33 | |
IgG | |
Affinity Purified | |
29 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q6PID6 | |
23548 | |
Synthetic peptides corresponding to TTC33(tetratricopeptide repeat domain 33) The peptide sequence was selected from the N terminal of TTC33. Peptide sequence DEGNWLHAIKRRKEILLEGCAEKSKQLKDEGASLAENKRYREAIQKWDEA. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title