Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC36 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen TTC36
Dilution Western Blot 1:500-1:2000, Immunohistochemistry -1:300-1:2000, Immunoprecipitation -1-5-ul/mg, Immunohistochemistry-Paraffin -1:300-1:2000
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121475
View Documents
Novus Biologicals
NBP31138350UL
50 μg
Each for $138.37
Add to cart
 
NB121474
View Documents
Novus Biologicals
NBP311383200UL
200 μg
Each for $320.71
Add to cart
 
Description

Description

TTC36 Polyclonal antibody specifically detects TTC36 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications

Specifications

TTC36
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Purified
RUO
PBS, pH7.0
143941
IgG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000, Immunohistochemistry -1:300-1:2000, Immunoprecipitation -1-5-ul/mg, Immunohistochemistry-Paraffin -1:300-1:2000
Unconjugated
Rabbit
Human
HBP21, tetratricopeptide repeat domain 36, tetratricopeptide repeat protein 36
E. coli-derived Human TTC36 fragment
Polyclonal
Antigen and protein A Affinity-purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit