Learn More
TTC37 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259165
Catalog No. NBP259165
Add to cart
Description
Description
TTC37 Polyclonal antibody specifically detects TTC37 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunofluorescence.
Specifications
Specifications
TTC37
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
tetratricopeptide repeat domain 37
Rabbit
IgG
150 ug
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot, ChIP assay, Immunofluorescence
Western Blot 1:1000, Chromatin Immunoprecipitation 1-5 ug, Immunofluorescence 1:100-1:500
Purified
TTC37
The exact immunogen is propietary information.
Protein G purified
RUO
Primary
9652