Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTC39C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566791
Description
Immunogen sequence: INMLLNNGFRE SDQLFKQYRN HSPLMSFGAS FVSFLNAMMT FEEEKMQLAC DDLKTTEKLC ESEEAGVIET IKNKIKKNVD VRKSAPSMVD RLQ Highest antigen sequence identity to the following orthologs - mouse 98%, rat 96%.
TTC39C is a protein coding gene.
Specifications
TTC39C | |
Polyclonal | |
Unconjugated | |
TTC39C | |
C18orf17; HsT2697; tetratricopeptide repeat domain 39C; tetratricopeptide repeat protein 39C; TPR repeat protein 39C; TTC39C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
125488 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N584 | |
TTC39C | |
Recombinant Human TTC39C. Recombinant protein control fragment (Product #RP-105366). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction