Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548264

Catalog No. PA548264

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TTC6
Polyclonal
PBS with 0.09% sodium azide
Q86TZ1
Tetratricopeptide repeat protein 6, TPR repeat protein 6, TTC6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
319089
Western Blot
Unconjugated
TTC6
Liquid
TTC6
KLH conjugated synthetic peptide between 410-438 amino acids from the C-terminal region of human TTC6.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit