Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC9B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548239

Catalog No. PA548239

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TTC9B
Polyclonal
PBS with 0.09% sodium azide
Q8N6N2, Q9D6E4
Tetratricopeptide repeat protein 9B, TPR repeat protein 9B, TTC9B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
148014, 73032
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TTC9B
Liquid
TTC9B
KLH conjugated synthetic peptide between 177-206 amino acids from the C-terminal region of human TTC9B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit