Learn More
TTF-1 (Thyroid Transcription Factor-1) Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ TTF-1 (Thyroid Transcription Factor-1) Ab-1, Mouse Monoclonal Antibody.
$770.00 - $1448.00
Specifications
Antigen | TTF-1 (Thyroid Transcription Factor-1) Ab-1 |
---|---|
Clone | 8G7G3/1 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Host Species | Mouse |
Catalog Number | Mfr. No. | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
MS699P
|
Epredia
MS699P |
Concentrated | Unconjugated | 1 mL |
Each for $1,448.00
|
|
|||
MS699P1
|
Epredia
MS699P1 |
Purified | Unconjugated | 500 μL |
Each for $842.00
|
|
|||
MS699RQ
|
Epredia
MS699RQ |
Ready-to-Use | Unconjugated | 12 mL |
Each for $770.00
|
|
|||
MS699B0
|
Epredia
MS699B0 |
Concentrated | Biotin | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS699B1
|
Epredia
MS699B1 |
Concentrated | Biotin | 500 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS699B
|
Epredia
MS699B |
Concentrated | Biotin | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS699P1ABX
|
Epredia
MS699P1ABX |
Concentrated | Unconjugated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS699PABX
|
Epredia
MS699PABX |
Concentrated | Unconjugated | 200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS699R1
|
Epredia
MS699R1 |
Prediluted | Unconjugated | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS699P0
|
Epredia
MS699P0 |
Purified | Unconjugated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
TTF-1(Thyroid transcription factor-1) is a member of the NKx2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate, and adrenal glands are unreactive. TTF-1 is detected in primary lung adenocarcinomas and small cell carcinomas and is absent in colon and breast carcinomas. Staining with TTF-1 antibody is useful for distinguishing between tumors of lung and non-lung origin.Host Species: Mouse
Clone: 8G7G3/1
Isotype: IgG1/κ
Species Reactivity: Human, Mouse and Rat. Shows a broad species reactivity.
Epitope: Not determined
Immunogen: Rat TTF-1 recombinant protein
Molecular Weight: 40kDa
Positive Control: MAD109 cells, normal thyroid or lung
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
TTF-1 (Thyroid Transcription Factor-1) Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
Mouse | |
Transcription Regulation | |
Primary |
8G7G3/1 | |
Monoclonal | |
IVD | |
Rat TTF-1 recombinant protein |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.