Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTF-1 / NKX2-1 Mouse anti-Human, Mouse, Rat, Biotin, Clone: 8G7G3/1 + NX2.1/690, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP247773B

Catalog No. NBP47773B


Add to cart

Description

Description

TTF-1 / NKX2-1 Monoclonal antibody specifically detects TTF-1 / NKX2-1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence.
Specifications

Specifications

TTF-1 / NKX2-1
8G7G3/1 + NX2.1/690
Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Purified
NKX2-1
Rat full length TTF-1 recombinant protein (8G7G3/1); Recombinant TTF-1 protein (NX2.1/690)
Protein A or G purified
Membrane Trafficking and Chaperones, Tumor Biomarkers
Primary
7080
Human, Mouse, Rat
Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Biotin
PBS with 0.05% Sodium Azide
benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2
Mouse
IgG
0.1 ml
Store at 4C in the dark.
Monoclonal
Recognizes a protein of 40kDa, identified as Thyroid transcription factor-1 (TTF-1). TTF-1 is a member of the NKx2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate, and adrenal glands are unreactive. Anti-TTF-1 is useful in differentiating primary adenocarcinoma of the lung from metastatic carcinomas originating in the breast, mediastinal germ cell tumors, and malignant mesothelioma. It can also be used to differentiate small cell lung carcinoma from lymphoid infiltrates.Loss of TTF-1 expression in non-small cell lung carcinoma has been associated with aggressive behavior of such neoplasms. TTF-1 reactivity is also seen in thyroid malignancies.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit