Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTF1 Rabbit anti-Human, Unconjugated, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation DH0070
Description
TTF1 Monoclonal antibody specifically detects TTF1 in Human samples. It is validated for IF.Specifications
TTF1 | |
Monoclonal | |
Unconjugated | |
TTF1 | |
Human recombinant TTF-1 | |
Primary | |
Human | |
Liquid |
Immunofluorescence | |
0.15 mg/mL | |
transcription termination factor, RNA polymerase I | |
Rabbit | |
100 μL | |
7270 | |
Store at -20°C. Do not freeze. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction