Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517286
Description
It is not recommended to aliquot this antibody.
TTK is a serine/threonine kinase closely related to SPK1, PIM 1 and CDC2 serine/threonine kinases. TTK is a dual-specificity kinase, which can also phosphorylate tyrosine. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. TTK encodes an alpha isoform of the constant regulatory subunit A. Alternatively spliced transcript variants have been described.
Specifications
TTK | |
Polyclonal | |
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
P33981 | |
TTK | |
Synthetic peptide corresponding to residues surrounding Glu137 of human TTK. | |
100 μL | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
TTK | |
AL022661; cancer/testis antigen 96; CT96; Dual specificity protein kinase TTK; esk; Esk1; monopolar spindle 1 kinase; monopolar spindle 1-like 1; MPH1; MPS1; MPS1L1; OTTHUMP00000016778; OTTHUMP00000220743; phosphotyrosine picked threonine kinase; phosphotyrosine picked threonine-protein kinase; PYT; TTK; Ttk protein kinase | |
Rabbit | |
Affinity chromatography | |
RUO | |
7272 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction