Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTLL12 Mouse anti-Human, Clone: OTI13H9, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA500775

Catalog No. 50-167-1917


Add to cart

Description

Description

TTLL12 (tubulin tyrosine ligase-like family, member 12) is a 644 amino acid protein that contains one TTL domain. Of the fourteen members of the TTLL family that modify tubulin, TTLL12 is the least characterized member. TTLL12 is highly expressed in a multitude of metastatic prostate cancer cell lines, therefore, it is considered a target for tumor therapy. Overexpression of TTLL12 is suggested to alter chromosome ploidy, whereas downregulation of TTLL12 influence several post-translational modifications of tubulin.
Specifications

Specifications

TTLL12
Monoclonal
0.32 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q14166
dJ526I14.2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23170
Immunohistochemistry (Paraffin), Western Blot
OTI13H9
Unconjugated
TTLL12
Liquid
TTLL12
Full length human recombit protein of human TTLL12 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit