Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTLL13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548213

Catalog No. PA548213

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Polyglutamylase which preferentially modifies alpha- tubulin. Involved in the side-chain elongation step of the polyglutamylation reaction rather than in the initiation step.
Specifications

Specifications

TTLL13
Polyclonal
PBS with 0.09% sodium azide
A6NNM8
Tubulin polyglutamylase TTLL13, 6---, Tubulin--tyrosine ligase-like protein 13, TTLL13
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
440307
Flow Cytometry, Western Blot
Unconjugated
TTLL13P
Liquid
TTLL13P
KLH conjugated synthetic peptide between 461-490 amino acids from the Central region of human TTLL13.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit