Learn More
TTPA Mouse anti-Human, Unconjugated, Clone: 3E7D10, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 674311IG150UL
This item is not returnable.
View return policy
Description
The TTPA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TTPA. This antibody recognizes human, mouse, rat, pig antigen. The TTPA antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
TTPA | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
3E7D10 | |
Unconjugated | |
150 μL | |
7274 | |
IgG2b |