Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTR Goat anti-Rat, Polyclonal , Abnova™

Goat polyclonal antibody raised against rat Ttr.

Manufacturer:  Abnova Corporation PAB29710

Catalog No. 89-935-059

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

TTR
Unconjugated
Affinity Purified
Ttr
Recombinant protein corresponding to rat Ttr.
Antigen Affinity Purification
RUO
Primary
24856
ELISA
In PBS, pH 7.2 (0.09% sodium azide)
LR1, prealbumin
Goat
IgG
100μg
Store at 4°C. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Rat
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit