Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTR mutant (Y78F) Mouse anti-Human, Clone: AD7, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $450.00
Specifications
Antigen | TTR mutant (Y78F) |
---|---|
Clone | AD7 |
Concentration | 1 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABN230725
|
MilliporeSigma
MABN230725UG |
25 μg |
Each for $177.00
|
|
|||||
MABN2307100
|
MilliporeSigma
MABN2307100UG |
100 μg |
Each for $450.00
|
|
|||||
Description
TTR mutant (Y78F) Monoclonal antibody specifically detects TTR mutant (Y78F) in Human samples. It is validated for ELISA, Immunoprecipitation, Western Blotting.Specifications
TTR mutant (Y78F) | |
1 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG2b | |
Protein G purified |
AD7 | |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Recombinant human Transthyretin with Y78F mutation. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title