Learn More
Saint Gobain Performance Plastics TUB F2475 1/4 x 1/2 50FT/PK
Supplier:  Saint Gobain Performance Plastics ACX00019
Encompass_Preferred
Saint-Gobain Tygon High-Purity Pressure Tubing Overall Length 50 ft. Maximum Pressure 150 psi Autoclavable Autoclavable
Catalog No. 50-206-8983
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
View alternate product