Learn More
TUB1 Rat anti-Human, Yeast, Clone: YL1/2, Abnova™
Rat monoclonal antibody raised against yeast TUB1.
Manufacturer:  Abnova Corporation MAB12664
Catalog No. 89-935-060
Add to cart
Description
Description
Specifications
Specifications
TUB1
YL1/2
Rat monoclonal antibody raised against yeast TUB1.
In PBS, pH 7.4 (0.09% sodium azide).
TUB1
Yeast TUB1
Affinity purification
RUO
Primary
854889
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Radioimmune Assays, Western Blot
Unconjugated
ELISA (1:100-1:1000) Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunoprecipitation Radioimmunoassay Western Blot The optimal working dilution should be determine
Liquid
Rat
IgG2a
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Yeast