Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBAL3 Mouse anti-Human, Clone: OTI1D7, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503924

Catalog No. 50-167-4702


Add to cart

Description

Description

Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.
Specifications

Specifications

TUBAL3
Monoclonal
0.93 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
A6NHL2
tubulin alpha like 3
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79861
Flow Cytometry, Western Blot
OTI1D7
Unconjugated
TUBAL3
Liquid
TUBAL3
Human recombit protein fragment corresponding to amino acids 150-446 of human TUBAL3 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit