Learn More
Azzota Corp Tubastatin A DFS Item
Manufacturer:  Azzota Corp C1321
Tubastatin A is a potent and selective HDAC6 inhibitor. Tubastatin A is a potent HDAC6 inhibitor with an IC50 value of 15 nM. Please request if need more quantities
Catalog No. 50-161-3191