Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBB2B Mouse anti-Human, Clone: OTI0C3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526268

Catalog No. PIMA526268


Add to cart

Description

Description

MA1-82191 recognizes class III beta-tubulin whixh is reported to be restricted to neuronal tissue. Clone TU-20 has been used to investigate tumors of neuronal origin including neuroblastoma and ganglioneuroma. Class III beta tubulin has not been detected in tumors of non neuronal origin.
Specifications

Specifications

TUBB2B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BVA1
Tubulin beta-2B chain, TUBB2B
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
347733
Immunocytochemistry, Immunofluorescence, Western Blot
OTI0C3
Unconjugated
TUBB2B
Liquid
TUBB2B
Full length human recombit protein of TUBB2B produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit