Learn More
Wilmad Labglass TUBE DRYING 90DEG 1410
Manufacturer:  Wilmad Labglass ML4095600
Encompass_Preferred
Tube, Drying, Bent 90degree. Joint Size: [ST] 14/10 Inner. Drying tube used to protect moisture sensitive reactions from water vapor in atmosphere while permitting the reaction system to remain open to the atmosphere.
Catalog No. 50-803-116
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.