Learn More
Wilmad Labglass TUBE DRYING U-SHAPED 14/20
Manufacturer:  Wilmad Labglass ML1855700
Encompass_Preferred
Tube, Drying, U-Shaped. Joint Size: [ST] 14/20 Inner. Drying tube used to protect moisture sensitive reactions from water vapor in atmosphere while permitting the reaction system to remain open to the atmosphere.
Catalog No. 50-807-494
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.