Learn More
Wilmad Labglass TUBE DRYING U-SHAPED 29/42
Manufacturer:  Wilmad Labglass LG6790104
Encompass_Preferred
Tube, Drying, U-Shaped. Joint Size: [ST] 29/42 Inner. Drying tube used to protect moisture sensitive reactions from water vapor in atmosphere while permitting the reaction system to remain open to the atmosphere.
Catalog No. 50-807-628
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.