Learn More
Thermo Scientific™ Tube Revolver / Rotator, Includes US plug
Increase your lab’s efficiency with our variable speed tube rotator with a small footprint, flexible design, and simple display help increase flexiblity and effectiveness.
Manufacturer:  Thermo Scientific™ 8888100112N
Catalog No. 11676341FS
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Interchangeable rotisseries accommodate different tube sizesfor maximum flexiblity. Each paddle can be adjusted from 0-90 degrees.
  • Achieve more effective mixing with “oscillating” mode. Oscillation will occur at every 40 degrees in the rotation cycle.
  • Small footprint
  • Plug-and-play simplicity
  • Maintenance-free motor, quiet operation