Learn More
Wilmad Labglass TUBE SCHWARTZ 159MM 14/20
Manufacturer:  Wilmad Labglass LG2170102
Encompass_Preferred
Tube, Absorption or Drying, Schwartz. Joint Size: [ST] 14/40; Length: 150mm. Side Hose Connectors located at the mid point of each neck, and with a hole in each Stopper coinciding with bore in side tube.
Catalog No. 50-804-606
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.