Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Andwin ScientificSupplier Diversity Partner TUBE STRP NEPTUNE 0.2ML 125/PK DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner 89092050
The 0.2mL 8-Tube Strips in assorted colors are manufactured out of high quality virgin polypropylene and are pre-sterile. This product is ideal for heat transfer due to its microthin walls. Caps for these tubes are sold separately.
Catalog No. 50-112-6034