Learn More
Saint Gobain Performance Plastics Tube Tygon 1/8 x 1/4 50FT/PK
Supplier:  Saint Gobain Performance Plastics AAA00007
Encompass_Preferred
Saint-Gobain Tygon S3 Clear Tubing Formula B-44-4X Diameter Inner 0.12 in. 3.2 mm Diameter Outer 1/4 in. 6.4 mm Autoclavable Autoclavable Certifications/Compliance FDA 21 CFR 175.300 3-A and NSF Standard 51 Hardness 65 (Shore A) Max. Operating Temperature 165 deg.F 79 deg.C
Catalog No. 50-206-8904
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.