Learn More
Aviva Systems Biology Corp TUBE1 Antibody - middle region (ARP56868_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56868_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TUBE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TUBE1. Alternate names: FLJ22589, TUBE, dJ142L7.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62018