Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBE1 Mouse anti-Human, Clone: 7G3B2, Abnova™

Mouse monoclonal antibody raised against partial recombinant human TUBE1.

Manufacturer:  Abnova Corporation MAB12240

Catalog No. 89-935-064


Add to cart

Description

Description

This gene encodes a member of the tubulin superfamily. This protein localizes to the centriolar sub-distal appendages that are associated with the older of the two centrioles after centrosome duplication. This protein plays a central role in organization of the microtubules during centriole duplication. A pseudogene of this gene is found on chromosome 5

Specifications

Specifications

TUBE1
7G3B2
Mouse monoclonal antibody raised against partial recombinant human TUBE1.
In PBS (0.05% sodium azide)
FLJ22589/TUBE/dJ142L7.2
Mouse
IgG1
RUO
Primary
51175
ELISA, Flow Cytometry, Western Blot
Unconjugated
ELISA (1:10000) Flow Cytometry (1:200-1:400) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Liquid
TUBE1
Recombinant protein corresponding to amino acids 314-472 of human TUBE1.
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit