Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBGCP2 Mouse anti-Human, Mouse, Rat, Porcine, Alexa Fluor™ 350, Clone: GCP2-01, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221793AF350

Catalog No. NP221793A30


Add to cart

Description

Description

TUBGCP2 Monoclonal antibody specifically detects TUBGCP2 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

TUBGCP2
GCP2-01
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Conjugated, Purified
TUBGCP2
GST-fusion protein containing amino acids 2-194 of mouse GCP2.
Protein A purified
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody GCP2-01 recognizes gamma-tubulin complex component 2 (GCP2), a 95-103 kDa protein. It recognizes also phosphorylated GCP2.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
Gamma-ring complex protein 103 kDa, gamma-tubulin complex component 2, gamma-tubulin complex protein 2, GCP-2, GCP2hGrip103, h103p, hGCP2, hSpc97, MGC138162, SPBC97, Spc97p, Spindle pole body protein Spc97 homolog, tubulin, gamma complex associated protein 2
Mouse
IgG2b Kappa
0.1 mL
Primary
10844.0
Human, Mouse, Porcine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit