Learn More
TUBGCP2 Mouse anti-Human, Mouse, Rat, Porcine, Alexa Fluor™ 594, Clone: GCP2-01, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221793AF594
Catalog No. NP221793A94
Add to cart
Description
Description
TUBGCP2 Monoclonal antibody specifically detects TUBGCP2 in Human,Mouse,Rat,Porcine samples. It is validated for Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
TUBGCP2
GCP2-01
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gamma-ring complex protein 103 kDa, gamma-tubulin complex component 2, gamma-tubulin complex protein 2, GCP-2, GCP2hGrip103, h103p, hGCP2, hSpc97, MGC138162, SPBC97, Spc97p, Spindle pole body protein Spc97 homolog, tubulin, gamma complex associated protein 2
Mouse
IgG2b
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody GCP2-01 recognizes gamma-tubulin complex component 2 (GCP2), a 95-103 kDa protein. It recognizes also phosphorylated GCP2.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
TUBGCP2
GST-fusion protein containing amino acids 2-194 of mouse GCP2.
0.1 mL
Primary
10844
Human, Mouse, Porcine, Rat