Learn More
Cole-Parmer Flexelene SFX5-7N SFX Flexible Polyethylene Tubing, 5/16 x 7/16, 100' per pack  DFS Item
Manufacturer:  Cole-Parmer 9618006
Applications: Pharmaceutical, biotech, and general lab applications. Characteristics: Good chemical resistance. Ultra-low extractables. Heat-sealable. Translucent. Certifications: Meets USP Class VI requirements. Temperature range: -40 to 100°F (-40 to 37°C) Sterilize: Ethylene oxide or gamma irradiation.
Catalog No. 50-630-284