Learn More
Cole-Parmer Masterflex Transfer Tubing, PTFE, 11/16"ID x 3/4" OD; 25 ft  DFS Item
Manufacturer:  Cole-Parmer 0660550
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures. Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent. Certification: Resin is FDA-compliant (21 CFR 177.1550) Temperature range: -400 to 500°F (-240 to 260°C) Sterilize: Autoclave, ethylene oxide, or dry heat.
Catalog No. 50-634-387